ART® Non-Filter Reload System, Thermo Scientific
Supplier: Thermo Scientific
3501-RT 3501-05-RT 3502-05-RT 3522-RT 3521-RT 3522-05-RT 3521-05-RT 3552-RT 3551-05-RT 3772-RT 3771-RT 3771-05-RT 3102-RT 3101-RT 3102-05-RT 3101-05-RT
89413-196CS 1010.15 CAD
89413-196 89413-200 89413-202 89413-224 89413-226 89413-228 89413-230 89413-244 89413-250 89413-280 89413-282 89413-286 89413-304 89413-306 89413-308 89413-310
ART® Non-Filter Reload System, Thermo Scientific
Pipette Tips
ART® portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in the laboratory.
The space saving, stackable tip packaging allows for tips to pass through offset holes in lower insert. The easy to use, blister sleeve guide rails align tip inserts for smooth transfer into the rack. Supplied in sterile packaging, the outer sleeve protects tips from exterior contamination while refilling inserts. The environmentally-friendly tip packaging option reduces plastic waste. The rigid insert design provides reliable tip loading into multichannel devices. All tips are traceable, with lot number and description printed on every insert.
Renowned for reliable quality, the ART® series of pipette tips is now offering a complete selection of Barrier, Non-Filter, and Specialty tips. All formats deliver proven quality, performance, and reliability, backed by the highest Thermo Scientific standards and worldwide support.
Certifications: RNase-, DNase-, DNA-, ATP-, and pyrogen-free.
Ordering information: An empty rack is required for Reload Tower use.