Capto™ MMC Multimodal Chromatography Media
Supplier: Cytiva
17531702 17531710
CA97067-758EA 1515.46 CAD
CA97067-758 CA97067-760
Capto™ MMC Multimodal Chromatography Media
Protein Purification Protein Purification Resins
Capto MMC is a multimodal cation exchanger with a different selectivity, compared with traditional ion exchangers.
  • High dynamic binding capacity at high conductivity increases productivity
  • High volume throughput cuts process times
  • Smaller unit operations give cost-effective processing and reduce investments
  • Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
The ligand contains caroxylic group and thus its features partly resemble a weak cation exchanger, but for Capto MMC also several other types of interactions are involved, including hydrogen bonding and hydrophobic interactions. Capto MMC is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto MMC meets industrial demands with security of supply and comprehensive technical and regulatory support.
It can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges. Capto MMC is a multimodal cation exchanger, belonging to the Capto family of BioProcess media for fast, efficient and cost-effective protein purification. Capto MMC combines agarose base matrix developments with innovative ligand chemistry. It is composed of a rigid, high-flow agarose matrix and a novel multimodal ligand. The multimodal functionality gives Capto MMC a different selectivity, compared with traditional ion exchangers. It can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges at both high and low ionic strengths.