HiTrap Columns, Capto MMC, Cytiva

Supplier: Cytiva
11-0032-73 11-0032-75 29400465
CA95055-924LPK 799.9 CAD
CA95055-924L CA95056-220L CA76633-052
HiTrap Columns, Capto MMC, Cytiva
Chromatography Columns
HiTrap Capto MMC is prepacked with Capto MMC, a multimodal BioProcess ion exchange resin. The multimodal functionality gives a different selectivity compared with traditional ion exchangers and it binds proteins at high or low ionic strength.

  • High dynamic binding capacity at high conductivity
  • Selectivity allows binding in the presence of salt
  • High volume throughput
  • HiTrap Capto MMC can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges

The multimodal functionality gives a differentMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5 bar [0.5 MPa] (70 psi)
pH range 2 - 12
Storage 4...30 °C, 20% Ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR