Capto™ SP ImpRes Hydrophobic Interaction Chromatography Media
Supplier: Cytiva
17546810 17546802
CA97067-934EA 500.65 CAD
CA97067-934 CA97067-932
Capto™ SP ImpRes Hydrophobic Interaction Chromatography Media
Protein Purification Protein Purification Resins
Capto SP ImpRes is a strong cation exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification.
  • High-resolution intermediate purification and polishing media based on the well-established Capto platform with traditional ligands
  • Flexibl e process design due to large operational window of flow rates and bed heights
  • High throughput purifications easy to optimize and scale-up
  • Higher manufacturing productivity enables improved process economy
  • BioProcess media developed and supported for large-scale chromatography
  • Prepacked HiTrap columns for easy screening and small-scale purification
  • Security of supply and comprehensive regulatory support
Capto SP ImpRes is a strong cation exchanger for the high-throughput intermediate purification and polishing steps of a wide range of biomolecules. This chromatography media is part of an expanded high-resolution platform based on the high-flow agarose Capto product line. By combining the high-flow characteristics of Capto media with a small particle size, Capto SP ImpRes delivers excellent pressure-flow properties with impressive resolution. The ability to run at higher flow rates and higher bed heights also increases flexibility in process design. Capto SP ImpRes is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical proteins. Based on the well-established Capto base matrix and with traditional ionic group ligands, it provides many opportunities for improved productivity and straightforward process development. Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto SP ImpRes meets industrial demands with security of supply and comprehensive technical and regulatory support.