Capto™ S Ion Exchange Chromatography Media

Supplier: Cytiva

17544101 17544110
CA97067-888EA 771.85 CAD
CA97067-888 CA97067-890
Capto™ S Ion Exchange Chromatography Media
Protein Purification Protein Purification Resins

Capto™ S is a strong cation exchange bioprocessing medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification when high volume throughput is essential.


  • Strong cation exchanger for high-productivity capture and intermediate purification when high volume throughput is essential
  • High dynamic binding capacity at high flow raises productivity
  • High-volume throughput cuts process times
  • Cost-effective processing with smaller unit operations


Capto S withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime. Capto S is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Capto S meets industrial demands with security of supply and comprehensive technical and regulatory support.


Capto S is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a strong sulfonate (S) cation exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto S raises the productivity in industrial downstream processes. Capto S withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.

Order Now

SPECIFICATIONS

Ligand Sulfonate group
Environmentally Preferable
Matrix Highly cross-linked agarose with dextran surface extender
pH range 4 - 12
Total ionic capacity 0,11 - 0,14 mmol Na+/ml
Storage temperature 4...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR