CM Sephadex™ C-50 Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
17-0220-02 17-0220-01
CA95016-810LEA 5308.56 CAD
CA95016-810L CA95055-938L
CM Sephadex™ C-50 Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Sephadex ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix.
  • Weak cation exchanger suitable batch techniques
  • High binding capacities
  • Avoid using oxidising agents and dextranase
CM Sephadex A-50 is a weak cation exchanger based on the well documented and well proven Sephadex base matrix. Sephadex ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages. Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available. Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.
These groups are attached to glucose units in the matrix by stable ether linkages.