HiTrap SP HP Cation Exchange Chromatography Columns
Supplier: Cytiva
17-1151-01 17-1152-01 29-0513-24
CA95055-864LPK 543.88 CAD
CA95055-864L CA95056-076L CA11033-030
HiTrap SP HP Cation Exchange Chromatography Columns
Chromatography Columns
HiTrap SP HP is a strong cation exchange chromatography column for high-resolution, small-scale protein purification.
  • Packed with SP Sepharose High Performance strong cation exchange resin
  • Small (34 µm) bead size delivers high-performance, high-resolution purifications
  • Can be used alone or connected in series
  • Convenient HiTrap format: designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
With a bead size of 34 µm, SP Sepharose HP resin provides good resolution at a broad pH range, allowing selection of the most suitable pH for each application.
These columns packed with SP Sepharose High Performance resin are utilized in a variety of applications, including:
• Purification of cell extract during rapid purification of recombinant histones
• Purification of BpsOMP38 to research porin involvement in cephalosporin and carbapenem resistance of Burkholderia pseudomallei
• Use with an ÄKTA FPLC system forexpansion of hematopoietic stem cells by recombinant hoxb4