HiScreen Capto Phenyl (High Sub) Columns, Cytiva
Supplier: Cytiva
HiScreen™
28992472 28-9924-72
CA97067-836EA 483.14 CAD
CA97067-836 97067-836
HiScreen Capto Phenyl (High Sub) Columns, Cytiva
Chromatography Columns
Capto™ Phenyl (High sub) and Capto™ Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
  • Excellent for method optimization and parameter screening due to the 10 cm bed height
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same resin using the same flow velocity
Capto™ Phenyl (high sub) is an excellent choice for initial and intermediate step purifications requiring a chromatography resin with resin to high hydrophobicity. Performance and selectivity are similar to that of Phenyl Sepharose™ 6 Fast Flow (high sub). Higher flow rates can be used with Capto™ Phenyl (high sub) during equilibration, wash and regeneration.