HiTrap Columns, Octyl FF, Cytiva
Supplier: Cytiva
17-5196-01 17-1359-01
CA95055-822LPK 1307.67 CAD
CA95055-822L CA95056-262L
HiTrap Columns, Octyl FF, Cytiva
Chromatography Columns
HiTrap Octyl FF are prepacked Octyl Sepharose 4 Fast Flow columns for hydrophobic interaction chromatography (HIC).
  • Provide convenient and fast small-scale protein purifications
  • Octyl Sepharose 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
The columns are used for small-scale protein purification as well as for screening of binding and elution conditions.