HiTrap Columns, Butyl FF, Cytiva
Supplier: Cytiva
17-5197-01 17-1357-01
CA95055-824LPK 1392.95 CAD
CA95055-824L CA95056-260L
HiTrap Columns, Butyl FF, Cytiva
Chromatography Columns
HiTrap Butyl FF are prepacked Butyl Sepharose 4 Fast Flow columns for hydrophobic interaction chromatography (HIC).
  • For screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Provide convenient and fast small-scale protein purifications
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
The columns are used forMore Product Information