HiTrap Columns, Phenyl FF (Low Sub), Cytiva
Supplier: Cytiva
17-5194-01 17-1353-01
CA95055-820LPK 1397.42 CAD
CA95055-820L CA95056-256L
HiTrap Columns, Phenyl FF (Low Sub), Cytiva
Chromatography Columns
Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.
  • Provide convenient and fast small-scale protein purifications
  • Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity
  • Two levels of substitution on Phenyl Sepharose 6 Fast Flow help find optimal selectivity and binding capacity
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
Two levels of substitution on Phenyl Sepharose 6 Fast Flow help find optimal selectivity and binding capacity. HiTrap Phenyl FF (Low Sub) are prepacked Phenyl Sepharose 6 Fast Flow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elution conditions.