HiLoad Superdex Columns, 200 PG, Cytiva

Supplier: Cytiva
28989335 28989336
CA71002-670EA 9250.28 CAD
CA71002-670 CA71002-672
HiLoad Superdex Columns, 200 PG, Cytiva
Chromatography Columns
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component.

  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • 1/16" fittings for convenient connections to ÄKTA design systems

Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr <10000 (More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 3 bar [0.3 MPa] (42 psi)
pH range 3 - 12
Storage 4 to 30 °C, 20% Ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR