TALON® Superflow™ Affinity Chromatography Media, Cytiva
Supplier: Cytiva
28957499
28957502
CA71006-006EA
256.27
CAD
CA71006-006
CA71006-008
TALON® Superflow™ Affinity Chromatography Media, Cytiva
Protein Purification
Protein Purification Resins
Cobalt-based chromatography medium designed for purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC)
- Suitable when Ni²⁺ is not the optimal choice of metal ion
- Compatible with commonly used IMAC reagents
- Allows protein purification under native or denaturing conditions
- Used with prokaryotic and eukaryotic expression systems