HiTrap™ TALON® crude Columns

Supplier: Cytiva
28953766 28953767 28-9537-67 29-0485-65
CA71005-998EA 497.5 CAD
CA71005-998 CA71006-000 71006-000 CA11033-006
HiTrap™ TALON® crude Columns
Chromatography Columns
HiTrap TALON® crude 1 and 5 ml columns allow for rapid separations and easy scale-up with a minimum of sample preparation and equipment.

  • Columns prepacked with cobalt-based chromatography media designed for purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC)
  • Suitable for IMAC purification of histidine-tagged proteins when Ni²⁺ is not the optimal choice of metal ion
  • Compatible with commonly used IMAC reagents and appropriate for purifying proteins under native or denaturing conditions
  • Prepacked HiTrap
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5 bar [0.5 MPa] (70 psi)
pH range 3 - 12
Storage 4...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR