HiTrap IMAC FF Affinity Chromatography Columns, Cytiva

Supplier: Cytiva
17-0921-04 17-0921-02
CA95055-958LPK 1579.49 CAD
CA95055-958L CA95056-228L
HiTrap IMAC FF Affinity Chromatography Columns, Cytiva
Chromatography Columns
HiTrap IMAC FF is prepacked with IMAC Sepharose 6 Fast Flow. The resin is charged with the metalion of your choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.

  • For optimizing purification of histidine-tagged proteins when Ni²⁺ is not the best choice of metal ion
  • Conveniently charge with your metal of choice
  • Optimal bead size for scaling up
  • High binding capacity

IMAC Sepharose FF is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5 bar [0.5 MPa] (70 psi)
pH range 3 - 12
Storage 4 to 30 °C, 20% Ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR