HiTrap Columns, Benzamidine FF (High Sub), Cytiva
Supplier: Cytiva
17-5143-02 17-5143-01 17-5144-01
CA95056-098LEA 669.69 CAD
CA95056-098L CA95056-198L CA95056-200L
HiTrap Columns, Benzamidine FF (High Sub), Cytiva
Chromatography Columns
HiTrap Benzamidine FF (high sub) are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
  • Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates
  • High binding capacity
  • It can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC system
HiTrap BenzamidineMore Product Information