HiTrap Columns, Heparin HP, Cytiva
Supplier: Cytiva
17-0407-03 17-0407-01 17-0406-01
CA95055-850LPK 3012.22 CAD
CA95055-850L CA95055-948L CA95056-036L
HiTrap Columns, Heparin HP, Cytiva
Chromatography Columns
HiTrap Heparin HP columns are designed for high-resolution purification of DNA-binding proteins, coagulation factors, and other plasma proteins.
  • Suitable for purification of antithrombin III, coagulation factors, and other plasma proteins
  • Useful for screening and optimization before scaling up to bulk resins such as Heparin 6 Fast Flow
  • High-resolution purification and elution in narrow peaks, minimizing the need for further concentration steps
  • Prepacked with Heparin Sepharose High Performance resin
  • HiTrap format compatible with syringes, pumps, and
More Product Information