HiTrap Columns, MabSelect Xtra, Cytiva
Supplier: Cytiva
28-4082-61 28-4082-58 28-4082-60
CA95055-972LPK 8356.72 CAD
CA95055-972L CA95056-162L CA95056-234L
HiTrap Columns, MabSelect Xtra, Cytiva
Chromatography Columns
HiTrap MabSelect Xtra columns are prepacked with MabSelect Xtra, a BioProcess medium for capture of monoclonal antbodies from large sample volumes.
  • Increased dynamic binding capacity with smaller particle size and greater porosity than MabSelect medium
  • MabSelect Xtra uses the same recombinant protein A ligand as MabSelect
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
  • Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs
The increased capacity of the medium is well suited for purification of Mabs from high-level expression feedstock.