HiTrap Columns, MabSelect, Cytiva

Supplier: Cytiva
28-4082-56 28-4082-53 28-4082-55
CA95055-970LPK 7769.06 CAD
CA95055-970L CA95056-160L CA95056-232L
HiTrap Columns, MabSelect, Cytiva
Chromatography Columns
HiTrap™ MabSelect™ columns are prepacked with MabSelect™, a Bioprocess medium for capuring of monoclonal antibodies(Mabs) from large sample volumes.

  • Specificity of binding to the Fc region of IgG is similar to that of native protein A, which allows one-step purification
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5 bar [0.5 MPa] (70 psi)
pH range 3 - 10
Storage 2 to 8 °C, 20% Ethanol (alternative storage solution 2% benzyl alcohol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR