Protein A Mag Sepharose Xtra
Supplier: Cytiva
28967056 28967062 28967062 28967056
CA89166-162EA 272.17 CAD
CA89166-162 CA89166-164 89166-164 89166-162
Protein A Mag Sepharose Xtra
Beads Magnetic Beads
Protein A Mag Sepharose Xtra magnetic beads are designed for rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants.
  • Visible and dense Sepharose based magnetic beads with protein A as the ligand make it easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation
  • Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized high capacity for efficient
More Product Information