Vivaspin Sample Concentrators, Cytiva
Supplier: Cytiva
28-9322-93 28-9322-94 28-9322-96 28-9323-17 28-9323-18 28-9323-19 28-9322-58 28-9322-40 28-9322-45 28-9322-47 28-9322-48 28-9322-57 28-9323-58 28-9323-59 28-9323-60 28-9323-61 28-9323-62 28-9323-63 28-9322-18 28-9322-23 28-9322-25 28-9322-35 28-9322-36 28-9322-37
CA95055-826LPK 579.07 CAD
CA95055-826L CA95055-828L CA95055-830L CA95055-832L CA95055-834L CA95055-836L CA95055-914L CA95055-988L CA95055-990L CA95055-992L CA95055-994L CA95055-996L CA95056-124L CA95056-126L CA95056-128L CA95056-130L CA95056-132L CA95056-134L CA95056-172L CA95056-174L CA95056-176L CA95056-178L CA95056-180L CA95056-182L
Vivaspin Sample Concentrators, Cytiva
Filters Centrifugal Filters
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
  • Dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • Vertical PES membrane minimises membrane blockage and tolerates high flow rates
  • Compatible pH range is from pH 1 to 9
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cutoff (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3000 to 100000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.