illustra ExoProStar, Cytiva
Supplier: Cytiva
US78220 US78210 US78211 US78212 US78225
CA71002-798EA 110.15 CAD
CA71002-798 CA71002-800 CA71002-802 CA71002-804 CA71002-806
illustra ExoProStar, Cytiva
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
illustra™ ExoProStar™ contains illustra™ Alkaline Phosphatase and Exonuclease 1, formulated to work together to remove unincorporated primers and nucleotides from amplification reactions in preparation for sequencing, cloning, genotyping or further DNA modification reactions.
  • Enzymes optimised to work together for high efficiency removal of unincorporated primers and nucleotides
  • Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction
  • Fast 30 minute protocol; complete heat inactivation of the enzymes within 15 minutes
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
illustra ExoProStar is our new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits!
The ExoProStar PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.