High Purity rNTPs, Cytiva
Supplier: Cytiva
27-1006-03
27-1200-04
27-2000-04
27-2025-01
27-2056-01
27-2066-01
27-2076-01
27-2086-01
CA95038-174LEA
845.91
CAD
CA95038-174L
CA95038-176L
CA95038-186L
CA95038-188L
CA95038-194L
CA95038-198L
CA95038-202L
CA95038-206L
High Purity rNTPs, Cytiva
Nucleic Acid Reagents
Nucleotides
Easy to use, time saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
- Each lot is tested for ribonuclease contamination to ensure success in the application
- Supplied as crystalline solid or in purified water as 100 mM solution (pH 7.5)
- Available either as individual products or in a convenient set containing ATP, CTP, GTP and UTP in separate tubes
Kit contents: Adenosine 5'-Triphosphate, 100 mM Solution (ATP); Cytidine 5'-Triphosphate, 100 mM Solution (CTP); Guanosine 5'-Triphosphate, 100 mM Solution (GTP); Uridine 5'-Triphosphate, 100 mM Solution (UTP)