High Purity dNTPs, Cytiva

Supplier: Cytiva

27-1850-04 27-1860-04 27-1870-04 27-1880-04
CA95038-178LEA 1448.33 CAD
CA95038-178L CA95038-180L CA95038-182L CA95038-184L
High Purity dNTPs, Cytiva
Nucleic Acid Reagents Nucleotides

dNTPs designed for applications requiring high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.


  • High purity dNTPs are functionally tested in long PCR to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with Thermo Sequenase DNA Polymerase
  • High purity (>98% triphosphate) confirmed by chromatographic analysis and 31P nuclear magnetic resonance (NMR) spectroscopy


Ordering information: Supplied as solids. For high purity solutions dNTPs, see the improved "illustra Solutions dNTPs".

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Purity ≥ 98% triphosphate
Storage conditions -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR