illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kits, Cytiva
Supplier: Cytiva
25660124 25660196 25660197
CA89233-916EA 574.21 CAD
CA89233-916 CA89233-918 CA89233-920
illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kits, Cytiva
Nucleic Acid Reagents Isothermal Amplification Reagents
illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 12 to 20 μg yield from nanogram amounts of DNA sample.
  • Representative amplification of the whole genome
  • Preformulated, predispensed, single dose, lyophilised cakes for rapid reaction set-up and greater reproducibility
  • Quick and simple automation friendly protocol; no thermal cycler required
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint
  • Yields high quality DNA (eg. for high-throughput genotyping and DNA archival)
Delivery information: illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit contains all of the components necessary for whole genome amplification by isothermal strand displacement amplification. The reaction mixture is provided as predispensed, single-dose, lyophilised cakes in either strips of 8 tubes, or 96-well plates.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more