DNA (Deoxyribonucleic acid) (from salmon testes), sonicated

Supplier: Cytiva

27-4565-01 27-4565-01
CA95040-286LEA 1292.17 CAD
95040-286 CA95040-286L
DNA (Deoxyribonucleic acid) (from salmon testes), sonicated
DNA (Deoxyribonucleic acid)

Genomic DNA sheared by sonication for use in hybridization experiments.


  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • Suitable for blocking non-specific binding sites in hybridization experiments to reduce background


Sonicated DNA from salmon testes.

Storage Temperature: Freezer
MDL Number: MFCD00130922
CAS Number: 9007-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR