DNA (Deoxyribonucleic acid) (from calf thymus), sonicated
Supplier: Cytiva
27-4563-01
27-4563-01
CA95040-284LEA
1560.5
CAD
CA95040-284L
95040-284
DNA (Deoxyribonucleic acid) (from calf thymus), sonicated
DNA (Deoxyribonucleic acid)
Genomic DNA sheared by sonication for use in hybridization experiments.
- Fragmented DNA with a size range from 200 to 6000 nucleotides
- Purified by phenol extraction and freeze dried
- Tested for absence of nuclease
DNA isolated from calf thymus is sonicated to fragment the DNA to an average size of 3000 bp.
Storage Temperature:
Freezer |
MDL Number:
MFCD00130922 CAS Number: 9007-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...