Amersham ECL Plex Fluorescent Rainbow Markers, Cytiva
Supplier: Cytiva
ECL™
RPN850E RPN851E RPN851E RPN850E
CA95040-080LEA 149.52 CAD
95040-080 CA95040-080L 95040-082 CA95040-082L
Amersham ECL Plex Fluorescent Rainbow Markers, Cytiva
Electrophoresis Reagents Molecular Weight Markers
Amersham™ ECL™ Plex™ fluorescent Rainbow™ markers are protein molecular weight markers optimised for use with the ECL™ Plex™ Western blotting detection system. They are designed to provide visible marker bands on gels and membranes, as well as fluorescent images using Cy3 and Cy5 channels.
  • Broad colour spectrum for easier band identification
  • Sharper, more intense bands on gels and blots
  • Ready-to-load formulations