Amersham™ CyDye™ Value Packs (Mono-Reactive Maleimide), Cytiva

Supplier: Cytiva

PA13131 PA15131 PA15636 PA15136 PA15130 PA15631
CA95017-516LEA 1058.61 CAD
CA95017-516L CA95017-518L CA95017-530L CA95017-534L CA95017-536L CA95056-538L
Amersham™ CyDye™ Value Packs (Mono-Reactive Maleimide), Cytiva
Protein Linking and Labeling Systems

Cy3 and Cy5 monofunctional maleimides are particularly suitable for the selective labeling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides and oligonucleotides.


  • Particularly effective for labeling antibodies
  • Large range of bulk packs available for maximum convenience and economy
  • Example protocols included with each dye pack


Also useful in antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application For labelling via thiol groups
For use in steady state fluorescence, FP and FRET applications
Solubility Soluble in cold water and hot water
Host
pH insensitive 3 pH Insensitive Min.; 10 pH Insensitive Max. (For 95017-518, 95017-536, 95017-534)
Storage conditions 2 - 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Shelf life 2 weeks (Estimated Shelf Life from Manufacture Date)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR