Thermoshakers for Microtubes and Microplates, Grant Instruments
Supplier: GRANT USA INC.
PHMT-PSC18 USA PHMT-PSC32 USA PHMT-PSC96 USA PHMT-PSC24 USA PHMTPSC24NUS
97025-570EA 5244.02 CAD
97025-570 89402-938 89402-940 89498-094 89498-096
Thermoshakers for Microtubes and Microplates, Grant Instruments
Shakers Shaking Incubators
The PHMT Series are compact, efficient and highly versatile thermoshakers, ideal for applications requiring microtube and microplate heating and shaking and suitable for use in cold rooms and incubators (operating temperature range 4 to 40ºC)
By combining the mixing operation with the incubation phase, reaction process times and operator workload are reduced and efficiency of many procedures is increased, resulting in a higher through put. The powerful motor operates extremely smoothly, quietly and consistently. Its 'soft' start prevents possible damage to samples from jolting. The PHMT-PSC18 block will accomodate up to twenty 0.5ml microtubes plus up to twelve 1.5ml microtubes.
The PHMT microtube thermoshaker has the following features: temperature setting range: ambient + 25 to 100ºC; temperature control range: ambient + 5 above ambient to 100ºC; uniformity: ± 0.1ºC; shaking speed: 250 to 1400 rpm; continuous or timed operation, with alarm buzzer and automatic switch-off facility; convenient interchangeable blocks; 2-line LCD display clearly indicates both set and actual values for temperature, shaking speed and time; compact and sturdy, with a low profile and small footprint; fits neatly into the workspace and provides years of reliable service.
The PHMT is ideal for DNA extraction, enzymic reactions and cellular biology techniques.
It is effectively three instruments in one: as a microtube and microplate thermoshaker; as a compact benchtop incubator without shaking; and as a microtube shaker without temperature control.
Certifications: CE approved.
Ordering information: Supplied complete with instructions, and 12V power supply. 2 Year warranty.