Ace-Thred Reducing Coupling, Ace Glass Incorporated
Supplier: Ace Glass
5842-05 5842-15 5842-46 5842-50 5842-10 5842-48
89052-656EA 107.86 CAD
89052-656 89052-660 89052-662 89052-666 89052-670 89052-680
Ace-Thred Reducing Coupling, Ace Glass Incorporated
Fittings and Connectors
These couplings are designed for threading columns together, or to secure end fittings with a single O-Ring seal for leak-tight engagement, with no significant size reduction in the I.D
One end is threaded for the next smallest diameter. Size listed refers to inside diameter of threads and corresponds to the column diameter. Fabricated from borosilicate glass.