PIG® DrainBlocker® Drain Covers with DuPont Elvaloy® Material, New Pig
Supplier: New Pig
PLR400 PLR401 PLR402 PLR403 PLR404 PLR405 PLR406 PLR412 PLR420 PLR430 PLR407 PLR408
89208-394EA 351.77 CAD
89208-394 89208-396 89208-398 89208-400 89208-402 89208-404 89208-406 89208-408 89208-410 89208-412 89208-414 89208-416
PIG® DrainBlocker® Drain Covers with DuPont Elvaloy® Material, New Pig
Spill Control Materials
Use cover for quick spill response or long-term, outdoor deployment without worrying about degradation
Cover withstands prolonged exposure to diesel fuel and gasoline, even when completely submerged. Ideal for drain protection where liquid transfers, washdowns or other leak- and spill-prone jobs are common. Eliminates the need to frequently deploy and remove a temporary cover or permanently seal your drains. High-visibility yellow top layer draws attention to spills for increased safety.
Vinyl composite top layer resists stretching, ripping and tearing. DuPont Elvaloy technology provides lifelong flexibility plus chemical, UV and infrared light resistance to vinyl top. Super-sealing urethane bottom layer conforms to uneven surfaces and tightly seals drain openings to block liquid