SeQuant® ZIC®-cHILIC HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
1.50670.0001 1.50656.0001 1.50657.0001 1.50658.0001 1.50659.0001 1.50660.0001 1.50661.0001 1.50662.0001 1.50669.0001 1.50656.0001 1.50657.0001 1.50658.0001 1.50659.0001 1.50660.0001 1.50661.0001 1.50662.0001 1.50669.0001 1.50670.0001
52428-710EA 2240.22 CAD
52428-710 52428-720 52428-722 52428-724 52428-726 52428-728 52428-730 52428-732 52428-738 52428-742 CA1.50656.0001 CA1.50657.0001 CA1.50658.0001 CA1.50659.0001 CA1.50660.0001 CA1.50661.0001 CA1.50662.0001 CA1.50666.0001 CA1.50669.0001 CA1.50670.0001
SeQuant® ZIC®-cHILIC HPLC Columns, MilliporeSigma
Chromatography Columns
SeQuant® ZIC®-cHILIC is designed for improved separation of polar hydrophilic compounds that have proved difficult to separate by reversed-phase HPLC or previous types of HILIC columns.
  • Complementary selectivity by reoriented charges in zwitterionic functional group
  • Perfect LC-MS compatibility due to low eluent buffer strengths and very low bleeding
  • High‐density bonded phosphorylcholine zwitterionic stationary phase
  • Very high stability and excellent batch-to-batch reproducibility
The orientation of its zwitterionic functional group provides more accessible positive charges, which results in a complementary selectivity with more retention particularly for organic acids and other negatively charged compounds. Especially suitable for LC-MS thanks to an extraordinary high column stability and a minimized column bleed.