Cleanert® COOH SPE Columns, Bonna-Agela
Supplier: Agela Technologies
CH0006 CH1001 CH2003 CH5003 CH5006
97017-986PK 853.52 CAD
97017-986 97017-990 97017-994 97017-996 97017-998
Cleanert® COOH SPE Columns, Bonna-Agela
Chromatography Columns
Cleanert® COOH SPE products consists of a propyl carboxylic acid on the inner silica surface.
  • Useful sorbent for quaternary ammonium salt
  • Consists of a propyl carboxylic acid
The pKa of the carboxylic acid group is approximately 3.8. It is a useful sorbent for quaternary ammonium salt and other strong cations. Based on silica.