PIG® Oil-Only Spill Kit in Extra-Large Response Chest, New Pig
Supplier: New Pig
KIT404-02 KIT404-03 KIT404-01
56620-368EA 4145.41 CAD
56620-368 56620-474 56620-476
PIG® Oil-Only Spill Kit in Extra-Large Response Chest, New Pig
Spill Control Materials
Seamless, polyethylene, lockable container keeps absorbents clean, dry and secure before use
  • Prepacked, compartmentalized container provides easy access to contents
  • Large, color-coded label makes identification fast and accurate
  • Temporary disposal bags help make cleanup easier
  • Tamperproof seals help prevent pilfering of spill response supplies
  • Four-way forklift entry for easy moving
The 10" no flat wheels roll easily over uneven surfaces. The 5" rubber wheels are well suited to smooth concrete or pavement. Also available without wheels. Not UN rated.
PIG Oil-Only Booms stop spreading spills; PIG Stat-Mat Pads and Pillows absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables.