NMR Tube Washer, Ace Glass Incorporated
Supplier: Ace Glass
2540-42 2540-15 2540-13 2540-31 2540-19 2540-25 2540-54 2540-29 2540-23 2540-12
89050-960EA 133.38 CAD
89050-960 89050-950 89050-962 89050-952 89050-964 89050-954 89050-966 89050-956 89050-968 89050-958
NMR Tube Washer, Ace Glass Incorporated
Tubes NMR Tubes
Rugged NMR tube washer designed with either Ace-Threds for ease and convenience or Ace-Thred and standard taper joints
The all-threaded version has a filter flask with a #15 Ace-Thred at the top for securing tube washer with a bushing, O-Ring, and #11 Ace-Thred side port with a removable 'Ace-Safe' hose connection for connecting to suction.
The threaded/jointed version has a filter flask with a standard taper 24/40 joint at top for connecting to inner standard taper joint at bottom of tube washer. It also features a serrated hose connection on the side for connecting to suction.
Washers accept a single 5 mm or 10 mm O.D. NMR tube. They features ¹/₈" O.D. PTFE tubing, rather than glass, to carry solvents from side reservoir to NMR tube in washing chamber. The tube to be washed is held in place in a #11 Ace-Thred with a PTFE bushing adapter (separate adapters for 5 mm and 10 mm tube). Solvent reservoir capacity is 55 mL and is connected to main chamber via #7 Ace-Threds and PTFE coupling.
Ordering information: Complete item consists of tube washer body, solvent reservoir, PTFE coupling, 5 mm and 10 mm tube bushing, filter flask, and ¹/₈" PTFE tubing.