ULTRA-TURRAX® T18 Digital Disperser, IKA® Works
Supplier: IKA Works
0003720001 3720000
89409-876EA 5504.18 CAD
89409-876 89409-878
ULTRA-TURRAX® T18 Digital Disperser, IKA® Works
Homogenizers Benchtop Homogenizers
Competitively priced dispersing instrument with digital speed display for volumes ranging from 1 to1500 mL (H2O)
Units are equipped with digital speed display, electronic speed control, error code display, and electronic overload protection. The stainless steel dispersing elements can be cleaned quickly and easily. Plastic disposable dispersing elements are available in two sizes. Quiet operation.
It offers a wide speed range, from 3000 to 25,000rpm, that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
Certifications: CE certified.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Dispersing elements, boss head clamps, stands, and other accessories are sold separately.