Chromolith® HighResolution RP-18e HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
1.52020.0001 1.52021.0001 1.52025.0001 1.52020.0001 1.52021.0001 1.52025.0001 1.52022.0001 1.52023.0001
EM1.52020.0001EA 1339.6 CAD
EM1.52020.0001 EM1.52021.0001 EM1.52025.0001 CA1.52020.0001 CA1.52021.0001 CA1.52025.0001 CA1.52022.0001 10811-854
Chromolith® HighResolution RP-18e HPLC Columns, MilliporeSigma
Chromatography Columns
The Chromolith® HighResolution columns offer higher efficiency and improved peak shape combined with lower back-pressure which is more than two times lower than any particulate packed column with the same dimensions.
  • High efficiencies at lower back pressure
  • Improved peak symmetry
  • Can be used for standard HPLC/UHPLC/LC-MS instruments
  • Back pressure 2 times lower compared to particulate packed columns
  • 30% longer column lifetime