Immersion Coolers and Flow-Through Cooler, JULABO
Supplier: JML SCIENCES INC CA
9650820.2 9650890.2 9650842.2 9650892.2 9655825.2 9650840.2 9650893.33 9650893.33 9650842.2 9650820 9650840 9650890 9650842 9650892 9655825
89421-944EA 10505.06 CAD
89421-944 89421-948 89421-950 89421-952 89421-954 89421-946 76003-780 CA76003-780 CA85002-876 CA89421-944 CA89421-946 CA89421-948 CA89421-950 CA89421-952 CA89421-954
Immersion Coolers and Flow-Through Cooler, JULABO
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Flow through cooler and immersion coolers for tap water and dry-ice replacement.
  • Replace tap water with FD200 flow-through cooler
  • Immersion coolers can replace dry ice
  • Units with Pt100 sensor can be set to a specific temperature
JULABO Immersion Coolers are primarily used for counter-cooling when connected to a heating circulator, or for rapidly cooling fluids down to low temperatures
The energy efficient units are easy to move around the laboratory with carrying handles; FT900/902/903 models have castors. The FT402, FT902 and FT903 models have anMore Product Information