BRAND macro™ Pipette Controller, BrandTech
Supplier: BrandTech
macro™
26203 26201 26200 26202
89410-178EA 270.96 CAD
89410-178 89410-176 89410-174 89410-180
BRAND macro™ Pipette Controller, BrandTech
Pipettes Pipette Controllers
macro™ pipette controller provides extremely smooth control.
  • Improved design now enables even finer control
  • Suitable for volumetric and serological pipettes from 0.1 to 200 ml
The entire macro™ pipette controller can be autoclaved at 121 °C (250 °F) for decontamination. The entire instrument can be disassembled for cleaning and the replacement of soiled filters or other parts.
Simply squeeze the suction bellows, which is sufficient for volumes up to 50 ml. Deflect the pipetting lever upward to aspirate, or downward to dispense. The further the deflection, the faster the aspiration and dispensing speeds. A new spring loaded pipetting lever allows even greater control with small volume pipettes. The proprietary 3 µm filter allows proper drain times for pipetting of Class A glassware, and helps prevent aqueous liquid penetration. A convenient blow out bulb allows accurate use of 'To Contain' pipettes. The suction bellows has been redesigned for even easier operation.
Ordering information: Each macro™ pipette controller is supplied in recyclable packaging with Operating Manual and spare 3 µm filter.