MRO Oil-Only Mat Pads, New Pig
Supplier: New Pig
MAT800 MAT801
56620-192PK 238.88 CAD
56620-192 56620-194
MRO Oil-Only Mat Pads, New Pig
Spill Control Materials
100% polypropylene absorbs petroleum-based liquids but not water
  • Pads are great for spill cleanup and smaller leak-and-drip applications
  • Rolls are great for catching overspray and machine leaks
Dimpled, 3-layer design quickly absorbs liquids to maximum capacity. Perforations let you tear off only what you need; reduces waste to save on disposal costs.
Order Now
SPECIFICATIONS
Environmentally Preferable