PIG® Dark Oil-Only Absorbent Boom, New Pig
Supplier: New Pig
BOM412 BOM413 BOM414 BOM415 BOM410 BOM411
56620-378PK 237.65 CAD
56620-378 56620-380 56620-400 56620-404 56620-422 56620-424
PIG® Dark Oil-Only Absorbent Boom, New Pig
Spill Control Materials
Dark color makes booms less noticeable when deployed on land or water
  • Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water
  • Strong, zinc-plated carbon steel attachment clips and rings allow you to link booms together for greater length
  • Can be incinerated after use to reduce waste or for fuels blending
Floats to confine and soak up spills on water; hugs ground for land-based spills. Spunbond polypropylene skin is UV resistant up to 4 months. Tough outer mesh is UV resistant for long-term outdoor use without degradation; lets fluids easily pass through to filler material. Rope running along length of boom withstands up to 400-lb. loads, adding strength for deployment and retrieval. Booms float at surface for easy retrieval, even when saturated