PIG® Oil-Only Absorbent Booms, New Pig
Supplier: New Pig
BOM304 BOM405 BOM406 BOM408 BOM407 BOM409
56620-014CS 300.74 CAD
56620-014 56620-016 56620-018 56620-376 56620-390 56620-402
PIG® Oil-Only Absorbent Booms, New Pig
Spill Control Materials
Floats to confine and soak up spills on water; hugs ground for land-based spills.
  • Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water
  • Strong, zinc-plated carbon steel attachment clips and rings allow you to link booms together for greater length
  • Bright white color makes booms easier to see in outdoor environments and clearly shows saturation level
  • Can be incinerated after use to reduce waste or for fuels blending
Tough outer mesh is UV resistant for long-term outdoor use without degradation; lets fluids easily pass through to filler material. Rope running along length of boom withstands up to 400-lb. loads, adding strength for deployment and retrieval. Spunbond polypropylene skin is UV resistant up to 12 months; meets NFPA 99 standards for static decay. Booms float at surface for easy retrieval, even when saturated.