PIG® Latching Drum Lid for Open-Head Steel Drums, New Pig
Supplier: New Pig
DRM659-BK DRM659-RD DRM659-YW DRM821-BK DRM821-YW DRM821-RD
56620-738EA 1449.48 CAD
56620-738 56620-740 56620-742 89032-424 89032-426 89032-428
PIG® Latching Drum Lid for Open-Head Steel Drums, New Pig
Lids Lids for Laboratory Vessels/Containers
Latching handle easily opens and closes with one hand.
  • Nitrile gasket and locking mechanism seal lid, reduces vapor emissions keeps drum contents dry and secure
  • Easy-access lid closes securely to help you meet haz-waste and closed-container regs
Hinge stops keep funnel lid upright when open. Durable, chemical-resistant powder coat helps ensure long service life. Lockable latch helps prevent unauthorized access. Hinge pin is made for easy lid removal without loosening band around drum. Hinged with one hand latch.
Accessories information: ReplacesMore Product Information