PIG® Oil-Only Spaghetti Booms, New Pig
Supplier: New Pig
BOM404 BOM204 BOM203 BOM202 BOM402 BOM403
56620-416PK 305.48 CAD
56620-416 56620-432 56620-450 56620-458 56620-482 56620-500
PIG® Oil-Only Spaghetti Booms, New Pig
Spill Control Materials
Mesh skin and spaghetti strip filler design is excellent for pulling in thick oil or for use in fast-moving waters
  • Absorbs and retains oils and oil-based liquids, including lubricants, fuels and crude oil, without absorbing a drop of water
  • Strong, zinc-plated carbon steel attachment clips and rings allow you to link booms together
  • Bright white color makes boom easier to see in outdoor environments and clearly shows saturation level
  • Can be incinerated after use to reduce waste or for fuels blending
Note: Not recommended for use on land. PIG Oil-OnlyMore Product Information