HighTech Series Refrigerated/Heating Circulators, JULABO
Supplier: JML SCIENCES INC CA
9400331.2 9400341.2 9400331 9400341
89421-722EA 17716.54 CAD
89421-722 89421-724 89421-726 89421-732 89421-734 89421-736 89421-738 89421-740 89421-742 89421-748 89421-750 89421-756 89421-758 CA89421-722 CA89421-724 CA89421-726 CA89421-732 CA89421-734 CA89421-736 CA89421-738 CA89421-740 CA89421-742 CA89421-748 CA89421-750 CA89421-756 CA89421-758
HighTech Series Refrigerated/Heating Circulators, JULABO
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
JULABO HighTech Refrigerated and Heating Circulators are ideal for temperature control applications to external systems and/or inside the bath tank.
HE models have a temperature range of –50 to 200 °C, TFT temperature display for actual and set point values and keypad for set point in put. The electronically controlled pressure/suction pump has an illuminated pump stage display. A Connection for a Pt100 sensor enables external measurement and control. The HE includes an integrated programmer with up to 10 steps, a timer function, RS232 interface for PC control, three point calibration andMore Product Information