PIG® Absorbent Drip Pan, New Pig
Supplier: New Pig
2410PP 2410PP-01
56620-090CS 352.91 CAD
56620-090 56620-516
PIG® Absorbent Drip Pan, New Pig
Spill Control Materials
Low-profile drip pan fits underneath drum faucets and other tight spaces; wide base won't tip over like unsturdy coffee cans.
  • Soft polypropylene cover catches drips without splashing and prevents highly absorbent polypropylene filler from spilling out
  • Highly absorbent polypropylene filler soaks up liquid on contact
  • Excellent for use under leaking drum faucets, spigots and flanges to absorb nuisance drips; keeps your floors clean and safe
Absorbs and retains oils, coolants, solvents and water.