PIG® Poly Drip Decks, New Pig
Supplier: New Pig
PAK527 PAK565 PAK566
89208-788EA 610.75 CAD
89208-788 89208-790 89208-792
PIG® Poly Drip Decks, New Pig
Spill Control Materials
Low-profile 5.75"H design is easy to load or unload; pumps and funnels are easy to access
  • Low-profile, 5.75"H design is easy to load or unload; pumps and funnels easy to access
  • Molded-in sump catches leaks, drips and spills, keeps storage areas clean and safe
  • Low-density polyethylene (LDPE) resists UV rays, rust, corrosion and most chemicals
Textured grating adds traction and lifts out for easy sump access. Translucent white sidewalls allow easy visual inspection.
Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe. Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals.