Pig Blue® Absorbent Mat Pad, New Pig
Supplier: New Pig
BLU100 BLU101
89132-764CS 189.6 CAD
89132-764 89132-768
Pig Blue® Absorbent Mat Pad, New Pig
Spill Control Materials
Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention. FiberFusion Technology fuses superabsorbent natural fibers with strong synthetic fibers for one high-performance absorbent. Stays together, even when picking up or shifting a fully saturated mat.
  • Absorbs liquid evenly for better wicking and retention
  • Super-absorbent natural fibers with strong synthetic fibers for high-performance absorbent
  • Stays together, even when picking up or shifting fully saturated mat
Contains 75% recycled fibers; ideal in situations where green products are desired or required
Heavyweight Pig Blue construction is super absorbent for your most demanding tasks and large-volume cleanups. Lightweight Pig Blue construction is ideal for your everyday tasks and cleanups.
Fire retardant for added safety; flame-tested in accordance with ASTM Standard E84-08. Absorbs oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids. Easy-tear perforations let you take only what you need, so you use less mat and save money.