SP Bel-Art Lab Companion™ Cabinet Style Vacuum Desiccators, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42400-4001 F42400-4011 F42400-4021 F42400-4031 F42400-4101 F42400-4111 F42400-4121 F42400-4131
30621-062EA 2497.44 CAD
30621-062 30621-064 30621-066 30621-068 30621-070 30621-072 30621-074 30621-076
SP Bel-Art Lab Companion™ Cabinet Style Vacuum Desiccators, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
These high-quality vacuum desiccators stay gastight allowing reliable dry storage or experiments over extended periods.
  • Hinged door design
  • Movable perforated shelves
  • Transparent polycarbonate construction
  • Amber-tined models protect light-sensitive samples
Able to maintain 29in.-Hg vacuum for over 72 hours at room temperature, they provide vacuum integrity resulting in lower maintenance and greater peace of mind
Constructed of transparent polycarbonate, these desiccators are durable, easy-to-clean, chemical-resistant, and allow quick visualizationMore Product Information